Assay Details
| Gene Symbol: SPAG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110261.1 | NP_001103731.1 | 379 | 173 | ||
| NM_001110262.1 | NP_001103732.1 | 359 | 153 | |||
| GenBank mRNA | AF466353.1 | - | 379 | 173 | ||
| AF466354.1 | - | 359 | 153 | |||
| AY528239.1 | - | 353 | 173 | |||
More Information
Set Membership: |
|

SPAG11B
Genomic Map