Assay Details
| Gene Symbol: MAMU-DRB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ601348.1 | - | 366 | 128 | ||
| AJ601350.1 | - | 366 | 128 | |||
| EU934774.1 | - | 366 | 128 | |||
| GQ471892.1 | - | 366 | 128 | |||
More Information
Set Membership: |
|

MAMU-DRB5
Genomic Map