Assay Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032961.1 | NP_001028133.1 | 592 | 77 | ||
| XM_015150994.2 | XP_015006480.1 | 1087 | 77 | |||
| GenBank mRNA | AJ622828.1 | - | 592 | 77 | ||
More Information
Set Membership: |
|

KLRF1
Genomic Map