Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042658.1 | NP_001036123.1 | 102 | 88 | ||
| GenBank mRNA | AY094979.1 | - | 102 | 88 | ||
| JU496101.1 | - | 361 | 88 | |||
More Information
Set Membership: |
|

CD1E
Genomic Map