Assay Details
| Gene Symbol: LYNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032944.1 | NP_001028116.1 | 121 | 70 | ||
| XM_015146094.2 | XP_015001580.2 | 353 | 70 | |||
| XM_015146095.2 | XP_015001581.2 | 606 | 70 | |||
| XM_015146096.2 | XP_015001582.2 | 923 | 70 | |||
| GenBank mRNA | AY422956.1 | - | 121 | 70 | ||
| JU336026.1 | - | 262 | 70 | |||
| JV047999.1 | - | 392 | 70 | |||
More Information
Set Membership: |
|

LYNX1
Genomic Map