Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195473.1 | NP_001182402.1 | 761 | 87 | ||
| GenBank mRNA | AY445037.2 | - | 711 | 87 | ||
| JU477783.1 | - | 776 | 87 | |||
More Information
Set Membership: |
|

CD2
Genomic Map