Assay Details
Gene Symbol: DEFB122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034196.1 | NP_001029368.1 | 74 | 66 | ||
GenBank mRNA | AY499409.1 | - | 74 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
