Assay Details
| Gene Symbol: CLCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032912.1 | NP_001028084.1 | 966 | 88 | ||
| GenBank mRNA | AY581306.1 | - | 966 | 88 | ||
| JU338536.1 | - | 902 | 88 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CLCA1
Genomic Map