Assay Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040219.1 | NP_001035309.1 | 165 | 104 | ||
| GenBank mRNA | AY635466.1 | - | 165 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP3A5| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040219.1 | NP_001035309.1 | 165 | 104 | ||
| GenBank mRNA | AY635466.1 | - | 165 | 104 | ||
Set Membership: |
|