Assay Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040219.1 | NP_001035309.1 | 677 | 140 | ||
| XM_028844765.1 | XP_028700598.1 | 1782 | 140 | |||
| GenBank mRNA | AY635466.1 | - | 677 | 140 | ||
More Information
Set Membership: |
|

CYP3A5
Genomic Map