Assay Details
| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032924.2 | NP_001028096.1 | 406 | 70 | ||
| GenBank mRNA | AY885223.1 | - | 411 | 70 | ||
| U51912.1 | - | 351 | 70 | |||
More Information
Set Membership: |
|

CST3
Genomic Map