Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032927.1 | NP_001028099.1 | 585 | 100 | ||
| GenBank mRNA | AY937248.1 | - | 585 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAEP| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032927.1 | NP_001028099.1 | 585 | 100 | ||
| GenBank mRNA | AY937248.1 | - | 585 | 100 | ||
Set Membership: |
|