Assay Details
| Gene Symbol: TIMM17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195660.1 | NP_001182589.1 | 215 | 81 | ||
| GenBank mRNA | CB308818.1 | - | 218 | 81 | ||
| CO644659.1 | - | 201 | 81 | |||
| DQ147924.1 | - | 118 | 81 | |||
| JU326829.1 | - | 93 | 81 | |||
| JU472778.1 | - | 203 | 81 | |||
| JV045909.1 | - | 219 | 81 | |||
| JV635773.1 | - | 227 | 81 | |||
More Information
Set Membership: |
|

TIMM17A
Genomic Map