Assay Details
| Gene Symbol: RABL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260670.2 | NP_001247599.1 | 746 | 68 | ||
| GenBank mRNA | DQ148034.1 | - | 244 | 68 | ||
| JU323199.1 | - | 829 | 68 | |||
| JU471428.1 | - | 754 | 68 | |||
| JV046060.1 | - | 746 | 68 | |||
| JV567305.1 | - | 892 | 68 | |||
| JV704339.1 | - | 973 | 68 | |||
| Gene Symbol: LOC114677631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847684.1 | XP_028703517.1 | 782 | 68 | ||
More Information
Set Membership: |
|

RABL2B
Genomic Map