Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114755.2 | NP_001108227.1 | 386 | 82 | ||
| NM_001348210.2 | NP_001335139.1 | 386 | 82 | |||
| GenBank mRNA | CO583610.1 | - | 310 | 82 | ||
| DQ148058.1 | - | 207 | 82 | |||
| JV047629.1 | - | 347 | 82 | |||
| JV474471.1 | - | 332 | 82 | |||
| JV689735.1 | - | 320 | 82 | |||

SELENOS
Genomic Map