Assay Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266703.1 | NP_001253632.1 | 321 | 74 | ||
| XM_028853381.1 | XP_028709214.1 | 276 | 74 | |||
| XM_028853382.1 | XP_028709215.1 | 198 | 74 | |||
| GenBank mRNA | DQ148090.1 | - | 283 | 74 | ||
| JU333277.1 | - | 325 | 74 | |||
| JV045160.1 | - | 321 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_028853382.1 transcripts.
This assay is in the 5'UTR region of the XM_028853382.1 transcripts.
Set Membership: |
|

PLPP4
Genomic Map