Assay Details
| Gene Symbol: LILRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040675.1 | NP_001035765.1 | 92 | 81 | ||
| GenBank mRNA | DQ155430.1 | - | 92 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LILRA3| Gene Symbol: LILRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040675.1 | NP_001035765.1 | 92 | 81 | ||
| GenBank mRNA | DQ155430.1 | - | 92 | 81 | ||
Set Membership: |
|