Assay Details
| Gene Symbol: ADGRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038662.1 | NP_001033751.1 | 95 | 73 | ||
| XM_028838330.1 | XP_028694163.1 | 249 | 73 | |||
| XM_028838331.1 | XP_028694164.1 | 249 | 73 | |||
| XM_028838333.1 | XP_028694166.1 | 252 | 73 | |||
| GenBank mRNA | DQ227271.1 | - | 95 | 73 | ||
| Gene Symbol: ADGRE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123131.2 | XP_014978617.1 | 453 | 73 | ||
| XM_015123132.2 | XP_014978618.1 | 453 | 73 | |||
| XM_015123133.2 | XP_014978619.1 | 452 | 73 | |||
| XM_015123134.2 | XP_014978620.1 | 453 | 73 | |||
| XM_028839871.1 | XP_028695704.1 | 453 | 73 | |||
| XM_028839872.1 | XP_028695705.1 | 453 | 73 | |||
| XM_028839873.1 | XP_028695706.1 | 287 | 73 | |||
More Information
Set Membership: |
|

ADGRE2
Genomic Map