Assay Details
| Gene Symbol: ADGRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038662.1 | NP_001033751.1 | 1214 | 79 | ||
| XM_028838330.1 | XP_028694163.1 | 1368 | 79 | |||
| XM_028838331.1 | XP_028694164.1 | 1368 | 79 | |||
| XM_028838333.1 | XP_028694166.1 | 1224 | 79 | |||
| GenBank mRNA | DQ227271.1 | - | 1214 | 79 | ||
| JU485336.1 | - | 155 | 79 | |||
More Information
Set Membership: |
|

ADGRE2
Genomic Map