Assay Details
| Gene Symbol: MSMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047131.1 | NP_001040596.1 | 136 | 92 | ||
| GenBank mRNA | M92161.1 | - | 136 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSMB| Gene Symbol: MSMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047131.1 | NP_001040596.1 | 136 | 92 | ||
| GenBank mRNA | M92161.1 | - | 136 | 92 | ||
Set Membership: |
|