Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032964.3 | NP_001028136.3 | 174 | 56 | ||
| XM_015131116.2 | XP_014986602.2 | 591 | 56 | |||
| XM_015131125.2 | XP_014986611.2 | 577 | 56 | |||
| XM_015131134.2 | XP_014986620.2 | 577 | 56 | |||
| XM_015131138.2 | XP_014986624.2 | 590 | 56 | |||
| XM_015131144.2 | XP_014986630.2 | 583 | 56 | |||
| GenBank mRNA | S70343.1 | - | 174 | 56 | ||
More Information
Set Membership: |
|

RHCE
Genomic Map