Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032964.3 | NP_001028136.3 | 656 | 57 | ||
| XM_015131109.2 | XP_014986595.2 | 735 | 57 | |||
| XM_015131116.2 | XP_014986602.2 | 1073 | 57 | |||
| XM_015131125.2 | XP_014986611.2 | 1059 | 57 | |||
| XM_015131134.2 | XP_014986620.2 | 1059 | 57 | |||
| XM_015131138.2 | XP_014986624.2 | 1072 | 57 | |||
| XM_015131144.2 | XP_014986630.2 | 1065 | 57 | |||
| GenBank mRNA | JV657960.1 | - | 369 | 57 | ||
| S70343.1 | - | 656 | 57 | |||
More Information
Set Membership: |
|

RHCE
Genomic Map