Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032964.3 | NP_001028136.3 | 1244 | 144 | ||
| GenBank mRNA | S70343.1 | - | 1244 | 144 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RHCE| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032964.3 | NP_001028136.3 | 1244 | 144 | ||
| GenBank mRNA | S70343.1 | - | 1244 | 144 | ||
Set Membership: |
|