Assay Details
| Gene Symbol: PRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047128.3 | NP_001040593.1 | 108 | |||
| XM_015135639.2 | XP_014991125.1 | 108 | ||||
| GenBank mRNA | U09018.1 | - | 612 | 108 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRL
Genomic Map