Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101733.1 | NP_001095203.1 | 138 | 107 | ||
| GenBank mRNA | JU334501.1 | - | 161 | 107 | ||
| JU475600.1 | - | 162 | 107 | |||
| X60236.1 | - | 138 | 107 | |||
More Information
Set Membership: |
|

LYZ
Genomic Map