Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101733.1 | NP_001095203.1 | 298 | 86 | ||
| GenBank mRNA | JU334501.1 | - | 321 | 86 | ||
| JU475600.1 | - | 322 | 86 | |||
| X60236.1 | - | 298 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYZ| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101733.1 | NP_001095203.1 | 298 | 86 | ||
| GenBank mRNA | JU334501.1 | - | 321 | 86 | ||
| JU475600.1 | - | 322 | 86 | |||
| X60236.1 | - | 298 | 86 | |||