Assay Details
| Gene Symbol: CYP2A23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040216.1 | NP_001035306.1 | 831 | 135 | ||
| GenBank mRNA | AY635459.1 | - | 831 | 135 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2A23| Gene Symbol: CYP2A23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040216.1 | NP_001035306.1 | 831 | 135 | ||
| GenBank mRNA | AY635459.1 | - | 831 | 135 | ||
Set Membership: |
|