Assay Details
| Gene Symbol: CYP2C75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040211.1 | NP_001035301.1 | 333 | 119 | ||
| GenBank mRNA | AY635463.1 | - | 333 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2C75| Gene Symbol: CYP2C75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040211.1 | NP_001035301.1 | 333 | 119 | ||
| GenBank mRNA | AY635463.1 | - | 333 | 119 | ||
Set Membership: |
|