Assay Details
| Gene Symbol: STRADB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265921.1 | NP_001252850.1 | 738 | 92 | ||
| XM_028830351.1 | XP_028686184.1 | 1063 | 92 | |||
| GenBank mRNA | AY680541.1 | - | 455 | 92 | ||
| JU335334.1 | - | 853 | 92 | |||
| JU476287.1 | - | 839 | 92 | |||
| JV047568.1 | - | 738 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STRADB
Genomic Map