Assay Details
Gene Symbol: HSP90B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195524.1 | NP_001182453.1 | 2301 | 132 | ||
GenBank mRNA | CO646465.1 | - | 86 | 132 | ||
DQ147987.1 | - | 263 | 132 | |||
JU336168.1 | - | 2332 | 132 | |||
JU336170.1 | - | 2332 | 132 | |||
JU336171.1 | - | 2332 | 132 | |||
JU477023.1 | - | 2376 | 132 | |||
JU477024.1 | - | 2363 | 132 | |||
JU477026.1 | - | 2363 | 132 | |||
JU477028.1 | - | 2363 | 132 | |||
JV048119.1 | - | 2317 | 132 | |||
JV048120.1 | - | 2317 | 132 | |||
JV048121.1 | - | 2365 | 132 | |||
JV048123.1 | - | 2317 | 132 | |||
JV722347.1 | - | 2352 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
