Assay Details
| Gene Symbol: LILRA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040674.1 | NP_001035764.1 | 1263 | 90 | ||
| GenBank mRNA | DQ155434.1 | - | 1263 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LILRA6| Gene Symbol: LILRA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040674.1 | NP_001035764.1 | 1263 | 90 | ||
| GenBank mRNA | DQ155434.1 | - | 1263 | 90 | ||
Set Membership: |
|