Assay Details
| Gene Symbol: LILRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040671.1 | NP_001035761.1 | 1720 | 95 | ||
| XM_028838341.1 | XP_028694174.1 | 1844 | 95 | |||
| GenBank mRNA | DQ155429.1 | - | 1720 | 95 | ||
More Information
Set Membership: |
|

LILRA2
Genomic Map