Assay Details
| Gene Symbol: DEFA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032923.4 | NP_001028095.3 | 56 | 65 | ||
| GenBank mRNA | AY859406.2 | - | 57 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001032923.4 transcripts.
Set Membership: |
|

DEFA4
Genomic Map