Assay Details
| Gene Symbol: TUBA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195391.1 | NP_001182320.1 | 1354 | 159 | ||
| XM_028828777.1 | XP_028684610.1 | 3678 | 159 | |||
| GenBank mRNA | AF141923.2 | - | 1237 | 159 | ||
| CN646945.1 | - | 598 | 159 | |||
| JV481386.1 | - | 213 | 159 | |||
| JV628316.1 | - | 1357 | 159 | |||
| Gene Symbol: TUBA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU477146.1 | - | 1376 | 159 | ||
| JV667159.1 | - | 1362 | 159 | |||
More Information
Set Membership: |
|

TUBA1A
Genomic Map