Assay Details
| Gene Symbol: CYP2C8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040210.1 | NP_001035300.1 | 1030 | 155 | ||
| XM_015147724.2 | XP_015003210.1 | 1140 | 155 | |||
| XM_015147725.2 | XP_015003211.1 | 1140 | 155 | |||
| XM_028853231.1 | XP_028709064.1 | 1140 | 155 | |||
| GenBank mRNA | AY635462.1 | - | 1030 | 155 | ||
More Information
Set Membership: |
|

CYP2C8
Genomic Map