Assay Details
| Gene Symbol: SLC35A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266759.1 | NP_001253688.1 | 387 | 83 | ||
| XM_015136880.2 | XP_014992366.1 | 848 | 83 | |||
| XM_028846675.1 | XP_028702508.1 | 184 | 83 | |||
| GenBank mRNA | JU332536.1 | - | 412 | 83 | ||
| JU474012.1 | - | 367 | 83 | |||
| JV045939.1 | - | 387 | 83 | |||
| JV705334.1 | - | 370 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC35A1
Genomic Map