Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266304.1 | NP_001253233.1 | 845 | 103 | ||
| XM_015139970.2 | XP_014995456.1 | 999 | 103 | |||
| XM_015139971.2 | XP_014995457.1 | 937 | 103 | |||
| XM_015139972.2 | XP_014995458.1 | 748 | 103 | |||
| XM_028849147.1 | XP_028704980.1 | 1098 | 103 | |||
| XM_028849148.1 | XP_028704981.1 | 1038 | 103 | |||
| XM_028849149.1 | XP_028704982.1 | 913 | 103 | |||
| GenBank mRNA | JU332550.1 | - | 997 | 103 | ||
| JU332551.1 | - | 757 | 103 | |||
| JU474022.1 | - | 845 | 103 | |||
| JU474023.1 | - | 809 | 103 | |||
| JV045951.1 | - | 751 | 103 | |||
| JV705375.1 | - | 642 | 103 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAIP1
Genomic Map