Assay Details
| Gene Symbol: ATP5PO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195583.1 | NP_001182512.1 | 352 | 82 | ||
| GenBank mRNA | CD766961.1 | - | 349 | 82 | ||
| CD767226.1 | - | 366 | 82 | |||
| CO644743.1 | - | 222 | 82 | |||
| JU335045.1 | - | 407 | 82 | |||
More Information
Set Membership: |
|

ATP5PO
Genomic Map