Assay Details
| Gene Symbol: CPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258081.1 | NP_001245010.1 | 1252 | 105 | ||
| GenBank mRNA | JU473276.1 | - | 1252 | 105 | ||
| JV044222.1 | - | 1415 | 105 | |||
| JV533763.1 | - | 1489 | 105 | |||
| JV571640.1 | - | 1317 | 105 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CPE
Genomic Map