Assay Details
| Gene Symbol: PNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193551.1 | NP_001180480.1 | 593 | 80 | ||
| GenBank mRNA | CN646333.1 | - | 593 | 80 | ||
| JU323241.1 | - | 627 | 80 | |||
| JU475742.1 | - | 547 | 80 | |||
| JV045361.1 | - | 546 | 80 | |||
More Information
Set Membership: |
|

PNP
Genomic Map