Assay Details
| Gene Symbol: RANBP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139869.2 | XP_014995355.2 | 1676 | 148 | ||
| XM_015139871.2 | XP_014995357.2 | 1751 | 148 | |||
| XM_028849385.1 | XP_028705218.1 | 1191 | 148 | |||
| GenBank mRNA | JU323448.1 | - | 1598 | 148 | ||
| JV704420.1 | - | 1575 | 148 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RANBP3L
Genomic Map