Assay Details
| Gene Symbol: PSMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266336.2 | NP_001253265.1 | 172 | 59 | ||
| GenBank mRNA | JU320587.1 | - | 172 | 59 | ||
| JU473016.1 | - | 180 | 59 | |||
| JV044003.1 | - | 175 | 59 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMC4
Genomic Map