Assay Details
| Gene Symbol: SIVA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261325.1 | NP_001248254.1 | 527 | 58 | ||
| XM_015144522.2 | XP_015000008.1 | 1061 | 58 | |||
| GenBank mRNA | JU323297.1 | - | 533 | 58 | ||
| JU472840.1 | - | 543 | 58 | |||
| JU472841.1 | - | 348 | 58 | |||
| JV045943.1 | - | 527 | 58 | |||
| JV638887.1 | - | 547 | 58 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SIVA1
Genomic Map