Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089651.4 | XP_001089651.1 | 316 | 70 | ||
| XM_001090810.4 | XP_001090810.1 | 316 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FGG
Genomic Map