Assay Details
| Gene Symbol: SERPINA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195350.2 | NP_001182279.2 | 700 | 96 | ||
| XM_015144281.2 | XP_014999767.2 | 759 | 96 | |||
| XM_015144282.2 | XP_014999768.2 | 717 | 96 | |||
| GenBank mRNA | CO583222.1 | - | 704 | 96 | ||
| CO583829.1 | - | 467 | 96 | |||
| JU332168.1 | - | 658 | 96 | |||
| JV641848.1 | - | 698 | 96 | |||
More Information
Set Membership: |
|

SERPINA3
Genomic Map