Assay Details
| Gene Symbol: HSD17B11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257582.1 | NP_001244511.1 | 508 | 134 | ||
| XM_015138742.2 | XP_014994228.2 | 795 | 134 | |||
| GenBank mRNA | JU334817.1 | - | 508 | 134 | ||
| JU475845.1 | - | 428 | 134 | |||
| JV047260.1 | - | 406 | 134 | |||
| JV636197.1 | - | 458 | 134 | |||
More Information
Set Membership: |
|

HSD17B11
Genomic Map