Assay Details
| Gene Symbol: APOA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834006.1 | XP_028689839.1 | 433 | 58 | ||
| GenBank mRNA | JV059699.1 | - | 148 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APOA1| Gene Symbol: APOA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834006.1 | XP_028689839.1 | 433 | 58 | ||
| GenBank mRNA | JV059699.1 | - | 148 | 58 | ||
Set Membership: |
|