Assay Details
| Gene Symbol: LOC706568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115567.2 | XP_014971053.1 | 927 | 75 | ||
| XM_015115568.2 | XP_014971054.1 | 927 | 75 | |||
| GenBank mRNA | JU325009.1 | - | 264 | 75 | ||
More Information
Set Membership: |
|

LOC706568
Genomic Map