Assay Details
| Gene Symbol: BPHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135440.2 | XP_014990926.1 | 420 | 67 | ||
| XM_015135441.2 | XP_014990927.1 | 420 | 67 | |||
| XM_028847874.1 | XP_028703707.1 | 442 | 67 | |||
| XM_028847875.1 | XP_028703708.1 | 898 | 67 | |||
| XM_028847876.1 | XP_028703709.1 | 751 | 67 | |||
| GenBank mRNA | JU332216.1 | - | 428 | 67 | ||
| JU472699.1 | - | 389 | 67 | |||
| JV069499.1 | - | 233 | 67 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BPHL
Genomic Map