Assay Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260869.1 | NP_001247798.1 | 868 | 106 | ||
| XM_015139973.2 | XP_014995459.1 | 949 | 106 | |||
| XM_015139974.2 | XP_014995460.1 | 949 | 106 | |||
| GenBank mRNA | JU321889.1 | - | 940 | 106 | ||
| JU321890.1 | - | 881 | 106 | |||
| JU475379.1 | - | 843 | 106 | |||
| JV045477.1 | - | 927 | 106 | |||
| JV045478.1 | - | 868 | 106 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HMGCS1
Genomic Map